missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-d3-N-nitrosoglycine, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | N-Nitrososarcosine-d3 |
| CAS | 1189871-94-0 |
| Molecular Formula | C3 2H3 H3 N2 O3 |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C3H6N2O3/c1-5(4-8)2-3(6)7/h2H2,1H3,(H,6,7)/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])N(CC(=O)O)N=O |
| IUPAC Name | 2-[nitroso(trideuteriomethyl)amino]acetic acid |
| Molecular Weight (g/mol) | 121.11 |
| Formula Weight | 121.0567 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction