missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-DL-alanine, 98%

Product Code. 15432509
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 15432509

Brand: Thermo Scientific Alfa Aesar H65287.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 600-21-5 | C4H9NO2 | 103.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 600-21-5
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00063136
InChI Key GDFAOVXKHJXLEI-UHFFFAOYSA-N
Synonym n-methyl-dl-alanine, 2-methylamino propanoic acid, h-n-me-dl-ala-oh, n-alpha-methyl-dl-alanine, n-me-dl-ala-oh, alanine, n-methyl-, l, dl-n-methylalanine, me-dl-ala-oh, n-methyl cdl-alanine, n-, a-methyl-dl-alanine
PubChem CID 4377
IUPAC Name 2-(methylamino)propanoic acid
SMILES CC(C(=O)O)NC

Specifications

Color White
Quantity 25 g
Formula Weight 103.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl-DL-alanine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.