missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl Formoterol Fumarate (2:1) (Mixture of Diastereomers), TRC

Product Code. 30477575
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477575 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477575 Supplier Toronto Research Chemicals Supplier No. TRCM29595025MG
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Methyl Formoterol Fumarate (2:1) (Mixture of Diastereomers)
CAS 1795133-97-9
Molecular Formula 2 C20 H26 N2 O4 . C4 H4 O4
Quantity 25 mg
Synonym N-[2-Hydroxy-5-[1-hydroxy-2-[[2-(4-methoxyphenyl)-1-methylethyl]methylamino]ethyl]phenyl]formamide 2-butenedioate (2:1), N-[2-Hydroxy-5-[1-hydroxy-2-[methyl-[2-(4-methoxyphenyl)-1-methylethyl]amino]ethyl]phenyl]formamide hemifumarate, N-[2-Hydroxy-5-[1-hydroxy-2-[[2-(4-methoxyphenyl)-1-methylethyl]methylamino]ethyl]phenyl]formamide hemifumarate, N-[2-Hydroxy-5-[1-hydroxy-2-[[1-(4-methoxyphenyl)propan-2-yl](methyl)amino]ethyl]phenyl]formamide fumarate (2:1), N-Methyl Formoterol hemifumarate, N-Methylformoterol hemifumarate, N-[2-Hydroxy-5-[1-hydroxy-2-[methyl[[2-(4-methoxyphenyl)-1-methylethyl]amino]ethyl]phenyl]formamide (2E)-2-Butenedioate Salt (2:1)
InChI Formula InChI=1S/2C20H26N2O4.C4H4O4/c2*1-14(10-15-4-7-17(26-3)8-5-15)22(2)12-20(25)16-6-9-19(24)18(11-16)21-13-23;5-3(6)1-2-4(7)8/h2*4-9,11,13-14,20,24-25H,10,12H2,1-3H3,(H,21,23);1-2H,(H,5,6)(H,7,8)/b;;2-1+
SMILES COc1ccc(CC(C)N(C)CC(O)c2ccc(O)c(NC=O)c2)cc1.COc3ccc(CC(C)N(C)CC(O)c4ccc(O)c(NC=O)c4)cc3.OC(=O)\C=C\C(=O)O
IUPAC Name (E)-but-2-enedioic acid;N-[2-hydroxy-5-[1-hydroxy-2-[1-(4-methoxyphenyl)propan-2-yl-methylamino]ethyl]phenyl]formamide
Molecular Weight (g/mol) 832.935
Formula Weight 832.3895
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.