missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-L-glutamic Acid, TRC

Product Code. 30009456
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009456 100 mg 100mg
30004479 50 mg 50mg
30012508 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009456 Supplier Toronto Research Chemicals Supplier No. TRCN240115100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6753-62-4
Molecular Formula C6H11NO4
Molecular Weight (g/mol) 161.16
Synonym N-Methyl-L-glutamic Acid, (2S)-2-(Methylamino)pentanedioic Acid, N-Methylglutamic Acid;
SMILES CN[C@@H](CCC(=O)O)C(=O)O

Specifications

InChI Formula InChI=1S/C6H11NO4/c1-7-4(6(10)11)2-3-5(8)9/h4,7H,2-3H2,1H3,(H,8,9)(H,10,11)/t4-/m0/s1
Quantity 100 mg
Shipping Condition Room Temperature
Chemical Name or Material N-Methyl-L-glutamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.