missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-L-prolinol, 96%

Product Code. 15429088
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15429088 25 g 25g
15419088 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15429088 Supplier Thermo Scientific Alfa Aesar Supplier No. H62573.14
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 34381-71-0 | C6H13NO | 115.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34381-71-0
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.18
MDL Number MFCD00011727
InChI Key VCOJPHPOVDIRJK-UHFFFAOYNA-N
Synonym n-methyl-l-prolinol, s---1-methyl-2-pyrrolidinemethanol, n-methylprolinol, 2s-1-methylpyrrolidin-2-yl methanol, s-1-methylpyrrolidin-2-yl methanol, 2-pyrrolidinemethanol, 1-methyl-, 2s, 2s-1-methyl-2-pyrrolidinyl methanol, s---2-hydroxymethyl-1-methylpyrrolidine, 1-methyl-2-pyrrolidinyl methanol #, n-methyl prolinol
PubChem CID 643492
SMILES CN1CCCC1CO

Specifications

Refractive Index 1.469
Density 0.968
Boiling Point 67°C to 69°C (12mmHg)
Quantity 25 g
Flash Point 63°C (145°F)
Optical Rotation −49° (c=5 in Methanol)
Beilstein 79851
IUPAC Name [(2S)-1-methylpyrrolidin-2-yl]methanol
Formula Weight 115.17
Percent Purity 96%
Chemical Name or Material N-Methyl-L-prolinol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.