missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[(1-methyl-1H-imidazol-5-yl)methyl]amine, 97%, Thermo Scientific™

Product Code. p-3175519
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10195992 250 mg Amber Glass Bottle 250mg
10259672 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10195992 Supplier Thermo Scientific Maybridge Supplier No. CC23646CB
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 384821-19-6
Molecular Formula C6H11N3
Molecular Weight (g/mol) 125.175
MDL Number MFCD07772801
InChI Key YZCXLAHTHBVYGB-UHFFFAOYSA-N
Synonym n-methyl-n-1-methyl-1h-imidazol-5-yl methyl amine, methyl-3-methyl-3h-imidazol-4-ylmethyl-amine, 1h-imidazole-5-methanamine, n,1-dimethyl, n-methyl-1-1-methyl-1h-imidazol-5-yl methanamine, methyl 3-methylimidazol-4-yl methyl amine, 1-methyl-5-methylamino methyl-1h-imidazole, methyl 1-methyl-1h-imidazol-5-yl methyl amine, n-methyl-1-3-methylimidazol-4-yl methanamine, n,3-dimethyl-3h-imidazole-4-methanamine, 1h-imidazole-5-methanamine,n,1-dimethyl
PubChem CID 7164555
IUPAC Name N-methyl-1-(3-methylimidazol-4-yl)methanamine
SMILES CNCC1=CN=CN1C

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 125.17
Percent Purity 90%
Chemical Name or Material N-Methyl-N-[(1-methyl-1H-imidazol-5-yl)methyl]amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.