missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-(3-pyridin-3-ylbenzyl)amine, 97%, Thermo Scientific™

Product Code. 10411313
Change view
Click to view available options
Quantity:
250 mg
5 g
Unit Size:
250mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10411313 250 mg 250mg
10786735 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10411313 Supplier Thermo Scientific Maybridge Supplier No. CC39846CB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 852180-72-4
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.269
MDL Number MFCD07368550
InChI Key LZJRRTLTBULOHU-UHFFFAOYSA-N
Synonym n-methyl-n-3-pyridin-3-ylbenzyl amine, methyl 3-pyridin-3-yl phenyl methyl amine, n-methyl-n-3-pyridin-3-yl benzylamine, benzenemethanamine,n-methyl-3-3-pyridinyl, n-methyl-1-3-pyridin-3-yl phenyl methanamine, n-methyl-1-3-pyridin-3-ylphenyl methanamine, n-methyl-3-pyridin-3-yl benzylamine, methyl 3-3-pyridyl phenyl methyl amine, 3-3-methylamino methyl phenyl pyridine
PubChem CID 7060576
IUPAC Name N-methyl-1-(3-pyridin-3-ylphenyl)methanamine
SMILES CNCC1=CC=CC(=C1)C2=CN=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 198.27
Percent Purity 97%
Chemical Name or Material N-Methyl-N-(3-pyridin-3-ylbenzyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.