missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[4-(piperidin-1-ylmethyl)benzyl]amine dihydrochloride, Tech., Thermo Scientific™

Product Code. 10431503
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10431503 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10431503 Supplier Thermo Scientific Maybridge Supplier No. CC45596DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 859833-23-1
CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Molecular Formula C14H24Cl2N2
MDL Number MFCD07772864
Quantity 1 g
Synonym n-methyl-n-4-piperidin-1-ylmethyl benzyl amine dihydrochloride, n-methyl-1-4-piperidin-1-ylmethyl phenyl methanamine dihydrochloride, methyl 4-piperidin-1-ylmethyl phenyl methyl amine dihydrochloride, n-methyl-1-4-piperidin-1-ylmethyl phenyl-methanamine dihydrochloride, n-methyl-1-4-piperidin-1-yl methyl phenyl methanamine-hydrogen chloride 1/2
InChI Key VEBSTHXHQLTMJG-UHFFFAOYSA-N
SMILES Cl.Cl.CNCC1=CC=C(CN2CCCCC2)C=C1
IUPAC Name N-methyl-1-[4-(piperidin-1-ylmethyl)phenyl]methanamine;dihydrochloride
Molecular Weight (g/mol) 291.26
PubChem CID 18525780
Formula Weight 291.26
Percent Purity Tech.
Grade Technical
Chemical Name or Material N-methyl-N-[4-(piperidin-1-ylmethyl)benzyl]amine dihydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.