missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[(5-methylthien-2-yl)methyl]amine monohydrochloride, 97%, Thermo Scientific™

Product Code. 10692325
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10692325 250 mg Amber Glass Bottle 250mg
10327554 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10692325 Supplier Thermo Scientific Maybridge Supplier No. CC65096CB
 Restricted Item
Estimated Shipment: 16-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 912569-78-9
Molecular Formula C7H12ClNS
Molecular Weight (g/mol) 177.69
MDL Number MFCD07106798
InChI Key OQOUPFNJXCXICY-UHFFFAOYSA-N
Synonym n-methyl-1-5-methylthiophen-2-yl methanamine hydrochloride, methyl 5-methylthiophen-2-yl methyl amine hydrochloride, n-methyl-n-5-methylthien-2-yl methyl amine hydrochloride, n-methyl-1-5-methyl-2-thienyl methanamine hydrochloride, 2-methyl-5-methylamino methyl thiophene hydrochloride, n-methyl-n-5-methylthien-2-yl methyl amine monohydrochloride, aronis23825, methyl 5-methyl 2-thienyl methyl amine, chloride, 2-methyl-5-methylamino methyl thiophene hcl, 2-thiophenemethanamine,n,5-dimethyl-,hydrochloride 1:1
PubChem CID 17290686
IUPAC Name N-methyl-1-(5-methylthiophen-2-yl)methanamine;hydrochloride
SMILES CC1=CC=C(S1)CNC.Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 177.7
Percent Purity 97%
Chemical Name or Material N-Methyl-N-[(5-methylthien-2-yl)methyl]amine monohydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.