missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-nitroso-1-octadecylamine, TRC

Product Code. 30138189
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138189 250 mg 250mg
30010512 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138189 Supplier Toronto Research Chemicals Supplier No. TRCM325200250MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69112-94-3
Molecular Formula C19 H40 N2 O
Molecular Weight (g/mol) 312.53
Synonym N-Methyl-N-nitrosooctadecylamine, N-Methyl-N-nitroso-1-octadecanamine
IUPAC Name N-methyl-N-octadecylnitrous amide
SMILES CCCCCCCCCCCCCCCCCCN(C)N=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C19H40N2O/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-21(2)20-22/h3-19H2,1-2H3
Formula Weight 312.3141
Percent Purity >95%
Chemical Name or Material N-Methyl-N-nitroso-1-octadecylamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.