missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-nitroso-ethenamine, TRC

Product Code. 30138349
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138349 500 mg 500mg
30136656 250 mg 250mg
30041095 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138349 Supplier Toronto Research Chemicals Supplier No. TRCM340995500MG
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4549-40-0
Molecular Formula C3H6N2O
Molecular Weight (g/mol) 86.09
Synonym N-methyl-N-nitroso- (6CI,7CI,8CI) Vinylamine;Methylvinylnitrosamine;N-Methyl-N-nitrosovinylamine;N-Nitroso-N-methylvinylamine;N-Nitrosomethylvinylamine;NSC 137906;Nitrosomethylvinylamine
IUPAC Name N-ethenyl-N-methylnitrous amide
SMILES CN(C=C)N=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C3H6N2O/c1-3-5(2)4-6/h3H,1H2,2H3
Formula Weight 86.048
Percent Purity >95%
Chemical Name or Material N-Methyl-N-nitroso-ethenamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.