missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-(quinolin-6-ylmethyl)amine, 95%, Thermo Scientific™

Product Code. 10778045
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10778045 1 g 1g
10366844 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10778045 Supplier Thermo Scientific Maybridge Supplier No. CC04546DA
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 179873-36-0
Molecular Formula C11H12N2
Molecular Weight (g/mol) 172.23
MDL Number MFCD06738897
InChI Key IIPNTNDPIZNFRU-UHFFFAOYSA-N
Synonym n-methyl-n-quinolin-6-ylmethyl amine, n-methyl-1-quinolin-6-yl methanamine, methyl quinolin-6-ylmethyl amine, methyl-quinolin-6-ylmethyl-amine, n-methyl-n-6-quinolinylmethyl amine, 6-quinolinemethanamine, n-methyl, 6-quinolinemethanamine,n-methyl, n-methyl-1-6-quinolinyl methanamine, n-methyl-6-quinolinemethanamine, methyl 6-quinolylmethyl amine
PubChem CID 6483903
IUPAC Name N-methyl-1-quinolin-6-ylmethanamine
SMILES CNCC1=CC=C2N=CC=CC2=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 1 g
Formula Weight 172.23
Percent Purity 95%
Chemical Name or Material N-Methyl-N-(quinolin-6-ylmethyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.