missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%

Product Code. 11454514
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454514 100 g 100g
11434514 5 g 5g
11444514 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454514 Supplier Thermo Scientific Alfa Aesar Supplier No. A13141.22
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 24589-78-4 | C6H12F3NOSi | 199.25 g/mol

N-Methyl-N-(trimethylsilyl)trifluoroacetamide is used as a silylating reagent that forms volatile derivatives for GC-MS analysis. It acts as a pharmaceutical intermediate and metabolite of Famprofazone in humans. It is also used in the detection of steroid hormones 17-alfa-ethynylestradiol and estrone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methyl-N-(trimethylsilyl)trifluoroacetamide is used as a silylating reagent that forms volatile derivatives for GC-MS analysis. It acts as a pharmaceutical intermediate and metabolite of Famprofazone in humans. It is also used in the detection of steroid hormones 17-alfa-ethynylestradiol and estrone.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24589-78-4
Molecular Formula C6H12F3NOSi
Molecular Weight (g/mol) 199.25
MDL Number MFCD00000411
InChI Key MSPCIZMDDUQPGJ-UHFFFAOYSA-N
Synonym mstfa, n-methyl-n-trimethylsilyl trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide, n-methyl-n-trimethylsilyltrifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl, n-methyl-n-trimethylsilyl-trifluoroacetamide, acmc-209tfm
PubChem CID 32510
ChEBI CHEBI:85064
IUPAC Name 2,2,2-trifluoro-N-methyl-N-trimethylsilylacetamide
SMILES CN(C(=O)C(F)(F)F)[Si](C)(C)C

Specifications

Density 1.076
Boiling Point 130°C to 132°C
Flash Point 25°C (77°F)
Refractive Index 1.38
Quantity 100 g
UN Number UN1993
Beilstein 1941550
Sensitivity Moisture sensitive
Solubility Information Immiscible with water.
Formula Weight 199.25
Percent Purity 97%
Chemical Name or Material N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.