missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-o-phenylenediamine, Dihydrochloride, TRC

Product Code. 30469724
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30469724 50 g 50g
30465316 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30469724 Supplier Toronto Research Chemicals Supplier No. TRCM32589150G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25148-68-9
Molecular Formula C7 H10 N2 . 2 Cl H
Molecular Weight (g/mol) 195.09
Synonym Telmisartan Ksm Ii
IUPAC Name 2-N-methylbenzene-1,2-diamine;dihydrochloride
SMILES Cl.Cl.CNc1ccccc1N

Specifications

Quantity 50 g
InChI Formula InChI=1S/C7H10N2.2ClH/c1-9-7-5-3-2-4-6(7)8;;/h2-5,9H,8H2,1H3;2*1H
Formula Weight 194.0378
Chemical Name or Material N-Methyl-o-phenylenediamine, Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.