missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl Pemetrexed, TRC

Product Code. 30137839
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137839 25 mg 25mg
30041104 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137839 Supplier Toronto Research Chemicals Supplier No. TRCM32546525MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 869791-42-4
Molecular Formula C21 H23 N5 O6
Molecular Weight (g/mol) 441.44
Synonym L-Glutamic acid, N-[4-[2-(2-amino-4,7-dihydro-1-methyl-4-oxo-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-, N-[4-[2-(2-Amino-4,7-dihydro-1-methyl-4-oxo-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-L-glutamic acid, N-Methyl Pemetrexed, N-Methylpemetrexed, (2S)-2-[[4-[2-(2-Amino-1-methyl-4-oxo-4,7-dihydro-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioic acid, Pemetrexed Disodium Heptahydrate Imp. A (EP)
IUPAC Name (2S)-2-[[4-[2-(2-amino-1-methyl-4-oxo-7H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioic acid
SMILES CN1C(=NC(=O)c2c(CCc3ccc(cc3)C(=O)N[C@@H](CCC(=O)O)C(=O)O)c[nH]c12)N

Specifications

InChI Formula InChI=1S/C21H23N5O6/c1-26-17-16(19(30)25-21(26)22)13(10-23-17)7-4-11-2-5-12(6-3-11)18(29)24-14(20(31)32)8-9-15(27)28/h2-3,5-6,10,14,23H,4,7-9H2,1H3,(H,24,29)(H,27,28)(H,31,32)(H2,22,25,30)/t14-/m0/s1
Quantity 25 mg
Formula Weight 441.1648
Shipping Condition Room Temperature
Chemical Name or Material N-Methyl Pemetrexed
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.