missing translation for 'onlineSavingsMsg'
Learn More

N-Methylaniline, 98%

Product Code. 11333297
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11333297 2500 g 2.5kg
11313297 100 g 100g
11323297 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11333297 Supplier Thermo Scientific Alfa Aesar Supplier No. L13873.0E
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 100-61-8 | C7H9N | 107.16 g/mol

N-Methylaniline is used in the preparation of N,N'-dimethylsulfinyldianilin using triethylamine and thionyl chloride as reagents. It is also acts as a latent and coupling solvent. Further, it serves as an intermediate for dyes and agrochemicals. In addition to this, it is utilized as an antiknock agent, which plays an essential role to increase the octane number.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylaniline is used in the preparation of N,N′-dimethylsulfinyldianilin using triethylamine and thionyl chloride as reagents. It is also acts as a latent and coupling solvent. Further, it serves as an intermediate for dyes and agrochemicals. In addition to this, it is utilized as an antiknock agent, which plays an essential role to increase the octane number.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents, acids, acid anhydrides, acid chlorides and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-61-8
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
MDL Number MFCD00008283
InChI Key AFBPFSWMIHJQDM-UHFFFAOYSA-N
Synonym methylaniline, monomethylaniline, n-methyl aniline, benzenamine, n-methyl, methylphenylamine, n-methylbenzenamine, n-monomethylaniline, n-methylaminobenzene, methylamino benzene, anilinomethane
PubChem CID 7515
ChEBI CHEBI:15733
IUPAC Name N-methylaniline
SMILES CNC1=CC=CC=C1

Specifications

Melting Point -57°C
Density 0.989
Boiling Point 194°C to 197°C
Flash Point 86°C (186°F)
Refractive Index 1.571
Quantity 2500 g
UN Number UN2294
Beilstein 741982
Sensitivity Air Sensitive
Merck Index 14,6019
Solubility Information Immiscible with water.
Formula Weight 107.16
Percent Purity 98%
Chemical Name or Material N-Methylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.