missing translation for 'onlineSavingsMsg'
Learn More

N-Methylcaprolactam, 96%

Product Code. 11463368
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463368 100 g 100g
11453368 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11463368 Supplier Thermo Scientific Alfa Aesar Supplier No. A17776.22
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2556-73-2 | C7H13NO | 127.187 g/mol

N-Methylcaprolactam was used to study the inhibition mechanism of poly-[N-vinylcaprolactam] (PVCAP) and its related compounds against hydrate formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylcaprolactam was used to study the inhibition mechanism of poly-[N-vinylcaprolactam] (PVCAP) and its related compounds against hydrate formation.

Solubility
Fully miscible with water.

Notes
Hygroscopic, store away from moisture. Store in a tightly closed container in a cool, dry, well-ventilated area. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2556-73-2
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.187
MDL Number MFCD00003263
InChI Key ZWXPDGCFMMFNRW-UHFFFAOYSA-N
Synonym n-methylcaprolactam, 1-methylcaprolactam, n-methyl-.epsilon.-caprolactam, 2h-azepin-2-one, hexahydro-1-methyl, n-methyl-e-caprolactam, epsilon-caprolactam, n-methyl, 2h-azepin-2-one, hexahydro-n-methyl, hexahydro-n-methyl-2h-azepin-2-one, n-methyl-epsilon-caprolactam, n-methyl caprolactam
PubChem CID 17369
IUPAC Name 1-methylazepan-2-one
SMILES CN1CCCCCC1=O

Specifications

Density 0.991
Boiling Point 106°C to 108°C (6 mmHg)
Flash Point 103°C (217°F)
Refractive Index 1.484
Quantity 100 g
Sensitivity Hygroscopic
Solubility Information Fully miscible with water.
Formula Weight 127.18
Percent Purity 96%
Chemical Name or Material N-Methylcaprolactam

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.