missing translation for 'onlineSavingsMsg'
Learn More

N-Methyldiacetamide, 90+%

Product Code. 11499796
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499796 5 g 5g
11409806 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499796 Supplier Thermo Scientific Alfa Aesar Supplier No. B20291.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1113-68-4 | C5H9NO2 | 115.132 g/mol

It can be used to produce N-Methyl-N-[1-(trimethylsiloxy)vinyl]acetamid at the ambient temperature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used to produce N-Methyl-N-[1-(trimethylsiloxy)vinyl]acetamid at the ambient temperature.

Solubility
Fully miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1113-68-4
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00014968
InChI Key ZNQFZPCFVNOXJQ-UHFFFAOYSA-N
Synonym n-methyldiacetamide, acetamide, n-acetyl-n-methyl, diacetamide, n-methyl, unii-767ut09b3u, n,n-diacetylmethylamine, n-methyl diacetamide, methyldiacetylamine, nn-diacetylmethylamine, acmc-1btf5, n-methyl-n-acetylacetamide
PubChem CID 14214
IUPAC Name N-acetyl-N-methylacetamide
SMILES CC(=O)N(C)C(=O)C

Specifications

Density 1.06
Boiling Point 192°C to 194°C
Refractive Index 1.453
Quantity 5 g
Solubility Information Fully miscible in water.
Formula Weight 115.13
Percent Purity ≥90%
Chemical Name or Material N-Methyldiacetamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.