missing translation for 'onlineSavingsMsg'
Learn More

N-Methylethylenediamine, 95%

Product Code. 10774281 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10774281 5 g Glass bottle 5g
10697312 25 g Glass bottle 25g
10455115 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10774281 Supplier Thermo Scientific Acros Supplier No. 155560050
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 109-81-9 | C3H12N2 | 76.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-81-9
Molecular Formula C3H12N2
Molecular Weight (g/mol) 76.14
MDL Number MFCD00008165
InChI Key KFIGICHILYTCJF-UHFFFAOYSA-P
Synonym n-methylethylenediamine, 1,2-ethanediamine, n-methyl, 2-aminoethylmethylamine, n-methyldiaminoethane, n-methylethanediamine, 2-methylamino ethylamine, n-methyl-1,2-ethanediamine, n-methylethylidenediamine, ethylenediamine, n-methyl, n-methylethane-1,2-diamine
PubChem CID 8014
IUPAC Name N'-methylethane-1,2-diamine
SMILES C[NH2+]CC[NH3+]

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8400g/mL
Boiling Point 114.0°C to 116.0°C
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula CH3NHCH2CH2NH2
Refractive Index 1.4390 to 1.4420
Quantity 5 g
Beilstein 04, I, 415
Specific Gravity 0.84
Formula Weight 74.13
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material N-Methylethylenediamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Methylethylenediamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.