missing translation for 'onlineSavingsMsg'
Learn More

N-Methylformamide, 99%

Product Code. 11396706
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396706 1000 g 1kg
11376706 50 g 50g
11386706 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11396706 Supplier Thermo Scientific Alfa Aesar Supplier No. L03908.0B
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 123-39-7 | C2H5NO | 59.068 g/mol

N-Methylformamide is used to prepare methyl isocyanate. It is used as an extraction solvent for aromatic hydrocarbons and acts as a ligand in coordination chemistry. It is employed as a solvent in oil refineries and in aluminum electrolytic capacitors. Further, it is used as a reagent in organic synthesis. In addition to this, it is used as an investigational anticancer drug and an intermediate in the production of some pharmaceutical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylformamide is used to prepare methyl isocyanate. It is used as an extraction solvent for aromatic hydrocarbons and acts as a ligand in coordination chemistry. It is employed as a solvent in oil refineries and in aluminum electrolytic capacitors. Further, it is used as a reagent in organic synthesis. In addition to this, it is used as an investigational anticancer drug and an intermediate in the production of some pharmaceutical compounds.

Solubility
Miscible with water, acetone and ethanol. Immiscible with ether.

Notes
Incompatible with strong oxidizing agents, acids, bases and acid chlorides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-39-7
Molecular Formula C2H5NO
Molecular Weight (g/mol) 59.068
MDL Number MFCD00003280
InChI Key ATHHXGZTWNVVOU-UHFFFAOYSA-N
Synonym methylformamide, monomethylformamide, formamide, n-methyl, n-formylmethylamine, n-methyl-formamide, n-monomethylformamide, n-methyl formamide, hconhch3, unii-xpe4g7y986, formamide, methyl
PubChem CID 31254
ChEBI CHEBI:7438
IUPAC Name N-methylformamide
SMILES CNC=O

Specifications

Melting Point -4°C
Density 1.018
Boiling Point 181°C to 185°C
Flash Point 98°C (208°F)
Linear Formula HCONHCH3
Refractive Index 1.432
Quantity 1000 g
Beilstein 1098352
Merck Index 14,6076
Solubility Information Miscible with water,acetone and ethanol. Immiscible with ether.
Formula Weight 59.07
Percent Purity 99%
Chemical Name or Material N-Methylformamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Methylformamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: N-Methylformamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.