missing translation for 'onlineSavingsMsg'
Learn More

N-Methylformamide-d5, TRC

Product Code. 30479767
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30479767 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30479767 Supplier Toronto Research Chemicals Supplier No. TRCN30538210MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Methylformamide-D5
CAS 863653-47-8
Molecular Formula C2D5NO
Quantity 10 mg
Synonym Formamide-N,1-d2, N-(methyl-d3)- (9CI), Perdeutero-N-methylformamide, N-Methylformamide-D5, N-(Methyl-d3)formamide-N,1-d2, N-Methylformamide-d5
InChI Formula InChI=1S/C2H5NO/c1-3-2-4/h2H,1H3,(H,3,4)/i1D3,2D/hD
SMILES [2H]C(N([2H])C([2H])([2H])[2H])=O
IUPAC Name N-(methyl-d3)formamide-N,1-d2
Molecular Weight (g/mol) 64.1
Formula Weight 64.07
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.