missing translation for 'onlineSavingsMsg'
Learn More

N-Methylformamide, TRC

Product Code. 30134668
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134668 100 g 100g
30011426 50 g 50g
30011304 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134668 Supplier Toronto Research Chemicals Supplier No. TRCM305380100G
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-39-7
Molecular Formula C2H5NO
Molecular Weight (g/mol) 59.07
Synonym Methylformamide, Monomethylformamide, N-Methylformimidic Acid, N-Monomethylformamide, NSC 3051
IUPAC Name N-methylformamide
SMILES CNC=O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C2H5NO/c1-3-2-4/h2H,1H3,(H,3,4)
Formula Weight 59.0371
Chemical Name or Material N-Methylformamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.