missing translation for 'onlineSavingsMsg'
Learn More

N-Methylformanilide, 99%

Product Code. 11493774
Change view
Click to view available options
Quantity:
10,000 g
250 g
2500 g
500 g
Unit Size:
10000g
2500g
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493774 500 g 500g
11428730 250 g 250g
11403784 2500 g 2500g
11413784 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11493774 Supplier Thermo Scientific Alfa Aesar Supplier No. A11829.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 93-61-8 | C8H9NO | 135.166 g/mol

N-Methylformanilide is used as a formylating reagent for certain organometallics. It is in combination with phosphoryl chloride used for Vilsmeier-Haack reactions and in heterocycle syntheses. Further, it acts as a swelling agent in the dyeing process of meta-aramid fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylformanilide is used as a formylating reagent for certain organometallics. It is in combination with phosphoryl chloride used for Vilsmeier-Haack reactions and in heterocycle syntheses. Further, it acts as a swelling agent in the dyeing process of meta-aramid fibers.

Solubility
Immisible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-61-8
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.166
MDL Number MFCD00003283
InChI Key JIKUXBYRTXDNIY-UHFFFAOYSA-N
Synonym n-methylformanilide, formamide, n-methyl-n-phenyl, n-formyl-n-methylaniline, formanilide, n-methyl, methylphenylformamide, n-phenyl-n-methylformamide, n-methyl-n-formylaniline, n-methylformanilid, methyl phenyl formamide, n-methyl-n-phenyl-formamide
PubChem CID 66737
IUPAC Name N-methyl-N-phenylformamide
SMILES CN(C=O)C1=CC=CC=C1

Specifications

Melting Point 9°C to 13°C
Density 1.095
Boiling Point 242°C to 244°C
Flash Point 126°C (258°F)
Refractive Index 1.561
Quantity 500 g
Beilstein 636496
Solubility Information Immisible with water.
Formula Weight 135.17
Percent Purity 99%
Chemical Name or Material N-Methylformanilide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.