missing translation for 'onlineSavingsMsg'
Learn More

N-Methylformanilide, 99%

Product Code. 13498338 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
10 kg
Packaging:
Glass Bottle
Plastic drum
Unit Size:
100g
10kg
1kg
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13498338 10 kg Plastic drum 10kg
10695362 5 g Glass Bottle 5g
10133130 100 g Glass Bottle 100g
10542521 500 g Glass Bottle 500g
10123130 1 kg Glass Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 13498338 Supplier Thermo Scientific Acros Supplier No. 126810100
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 93-61-8 | C8H9NO | 135.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-61-8
MDL Number MFCD00003283
InChI Key JIKUXBYRTXDNIY-UHFFFAOYSA-N
Synonym n-methylformanilide, formamide, n-methyl-n-phenyl, n-formyl-n-methylaniline, formanilide, n-methyl, methylphenylformamide, n-phenyl-n-methylformamide, n-methyl-n-formylaniline, n-methylformanilid, methyl phenyl formamide, n-methyl-n-phenyl-formamide
PubChem CID 66737
IUPAC Name N-methyl-N-phenylformamide
SMILES CN(C=O)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 8.0°C to 13.0°C
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 243.0°C to 244.0°C
Flash Point 126°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic drum
Molecular Formula C8H9NO
Linear Formula HCON(CH3)C6H5
Refractive Index 1.5600 to 1.5620
Quantity 10 kg
Beilstein 12,234
Fieser 01,68
Specific Gravity 1.09
Solubility Information Solubility in water: immiscible.
Molecular Weight (g/mol) 135.17
Formula Weight 135.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-Methylformanilide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • N-Methylformanilide
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.