missing translation for 'onlineSavingsMsg'
Learn More

N-Methylisatoic anhydride, tech. 90%

Product Code. 15463257
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15463257 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15463257 Supplier Thermo Scientific Alfa Aesar Supplier No. B24099.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 10328-92-4 | C9H7NO3 | 177.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 10328-92-4
Melting Point ∼160°C (decomposition)
Molecular Formula C9H7NO3
MDL Number MFCD00006815
Quantity 100 g
Sensitivity Moisture sensitive
Synonym n-methylisatoic anhydride, 1-methyl-1h-benzo d 1,3 oxazine-2,4-dione, n-methyl ia, 1-methyl-2h-3,1-benzoxazine-2,4 1h-dione, 2h-3,1-benzoxazine-2,4 1h-dione, 1-methyl, n-methyl-n-carboxyanthranilic anhydride, methyl-n-carboxyanthranilic anhydride, 1-methylbenzo d 1,3-oxazine-2,4-dione, n-methyl isotoic anhydride, 1-methyl-2,4-dihydro-1h-3,1-benzoxazine-2,4-dione
InChI Key KJMRWDHBVCNLTQ-UHFFFAOYSA-N
SMILES CN1C2=CC=CC=C2C(=O)OC1=O
IUPAC Name 1-methyl-3,1-benzoxazine-2,4-dione
Molecular Weight (g/mol) 177.159
PubChem CID 25160
ChEBI CHEBI:59054
Formula Weight 177.16
Percent Purity 90%
Grade Technical
Chemical Name or Material N-Methylisatoic anhydride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.