missing translation for 'onlineSavingsMsg'
Learn More

N-Methylisobutylamine, 98+%

Product Code. 11360218
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11360218 5 g 5g
11370218 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11360218 Supplier Thermo Scientific Alfa Aesar Supplier No. 042947.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 625-43-4 | C5H13N | 87.166 g/mol

N-Methylisobutylamine is used as an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-43-4
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00015043
InChI Key QKYWADPCTHTJHQ-UHFFFAOYSA-N
Synonym n-methylisobutylamine, 1-propanamine, n,2-dimethyl, n,2-dimethylpropylamine, n-isobutylmethylamine, methyl 2-methylpropyl amine, isobutylmethylamine, methylisobutylamine, propylamine, n,2-dimethyl, n-isobutyl-n-methylamine, methyl-isobutyl-amin
PubChem CID 12249
IUPAC Name N,2-dimethylpropan-1-amine
SMILES CC(C)CNC

Specifications

Density 0.73 g/mL
Boiling Point 78°C
Odor Amine-like
Quantity 5 g
UN Number UN2733
Solubility Information Miscible with water.
Formula Weight 87.16
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material N-Methylisobutylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.