missing translation for 'onlineSavingsMsg'
Learn More

N-Methylmaleic hydrazide, 97%

Product Code. 11440047
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440047 5 g 5g
11450047 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440047 Supplier Thermo Scientific Alfa Aesar Supplier No. B20975.06
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5436-01-1 | C5H6N2O2 | 126.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5436-01-1
MDL Number MFCD00234108

Specifications

Melting Point 216°C to 219°C
Molecular Formula C10H13NO2
Quantity 5 g
Synonym 2-amino-2-phenylbutyric acid, 2-ammonio-2-phenylbutanoate, benzeneacetic acid, a-amino-a-ethyl, 1-2-amino-2-phenylbutyric acid, aminophenylbutyric acid, acmc-20ao5m, 4-sulfobenzoic anhydride, 2-ethyl-2-phenylglycine, 2-amino-2-phenulbutyric acid, 2-amino-2-phenyl-butyric acid
InChI Key UBXUDSPYIGPGGP-UHFFFAOYNA-N
SMILES CCC(C1=CC=CC=C1)(C(=O)O)N
IUPAC Name 2-amino-2-phenylbutanoic acid
Molecular Weight (g/mol) 179.22
PubChem CID 89317
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material N-Methylmaleic hydrazide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.