missing translation for 'onlineSavingsMsg'
Learn More

N-Methylmaleimide, TRC

Product Code. 30483029
Change view
Click to view available options
Quantity:
5 g
500 g
Unit Size:
500g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483029 500 g 500g
30476739 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30483029 Supplier Toronto Research Chemicals Supplier No. TRCM316800500G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-88-1
Molecular Formula C5H5NO2
Molecular Weight (g/mol) 111.1
Synonym 1-Methyl-1H-pyrrole-2,5-dione, 1-Methylpyrrole-2,5-dione, N-Methylmaleinimide, N-Methylpyrrole-2,5-dione, NSC 57594
IUPAC Name 1-methylpyrrole-2,5-dione
SMILES CN1C(=O)C=CC1=O

Specifications

Quantity 500 g
InChI Formula InChI=1S/C5H5NO2/c1-6-4(7)2-3-5(6)8/h2-3H,1H3
Formula Weight 111.032
Chemical Name or Material N-Methylmaleimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.