missing translation for 'onlineSavingsMsg'
Learn More

N-Methylphenethylamine, 99%

Product Code. 10367150 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10367150 5 g Glass bottle 5g
10572331 25 g Glass bottle 25g
10617294 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10367150 Supplier Thermo Scientific Acros Supplier No. 127450050
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 589-08-2 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-08-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00008291
InChI Key SASNBVQSOZSTPD-UHFFFAOYSA-N
Synonym n-methylphenethylamine, benzeneethanamine, n-methyl, methyl-phenethyl-amine, n-methyl-2-phenylethylamine, n-methyl-n-2-phenylethyl amine, 1-phenyl-2-methylaminoethane, phenethylamine, n-methyl, methyl 2-phenylethyl amine, 1-phenyl-2-methylamino-aethan, n-methyl-phenethylamine
PubChem CID 11503
IUPAC Name N-methyl-2-phenylethanamine
SMILES CNCCC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 203.0°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2CH2NHCH3
Refractive Index 1.5150 to 1.5170
Quantity 5 g
Beilstein 12, 1097
Specific Gravity 0.93
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in alcohol and ether
Formula Weight 135.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-Methylphenethylamine
Show More Show Less
compliance-icons
Product Identifier
  • N-Methylphenethylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.