missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpyridinium-4-carboxaldehyde benzenesulfonate hydrate, 97%

Product Code. 11318605
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318605 25 g 25g
11308605 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318605 Supplier Thermo Scientific Alfa Aesar Supplier No. L12072.14
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 82228-89-5 | C13H13NO4S | 279.31 g/mol

N-Methylpyridinium-4-carboxaldehyde benzenesulfonate hydrate is a useful reagent for the chemical transformation of primary amines to aldehydes and ketones under extremely mild conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylpyridinium-4-carboxaldehyde benzenesulfonate hydrate is a useful reagent for the chemical transformation of primary amines to aldehydes and ketones under extremely mild conditions.

Solubility
Sparingly soluble in water (0.27 g/L at 25°C).

Notes
Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature. Store under inert gas. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82228-89-5
Molecular Formula C13H13NO4S
Molecular Weight (g/mol) 279.31
MDL Number MFCD00043164
InChI Key HSVLGIFAXFDLMU-UHFFFAOYSA-M
Synonym 4-formyl-1-methylpyridinium benzenesulfonate, 4-formyl-1-methyl-pyridinium benzenesulfonate, 4-formyl-1-methylpyridin-1-ium benzenesulfonate, pyridinium, 4-formyl-1-methyl-, benzenesulfonate, benzenesulfonate; 1-methylpyridin-1-ium-4-carbaldehyde, 4-formyl-1-methyl pyridinium benzensulfonate, 4-formyl-l-methyl pyridinium benzensulfonate, 4-formyl-1-methylpyridinium benzenesulfo nate, benzenesulfonic acid, 1-methylpyridine-4-carbaldehyde
PubChem CID 9817127
IUPAC Name benzenesulfonate;1-methylpyridin-1-ium-4-carbaldehyde
SMILES C[N+]1=CC=C(C=O)C=C1.[O-]S(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point 98°C to 100°C
Quantity 25 g
Beilstein 5695963
Sensitivity Air Sensitive
Solubility Information Sparingly soluble in water (0.27g/L at 25°C).
Formula Weight 279.32 (Anhydrous)
Percent Purity 97%
Chemical Name or Material N-Methylpyridinium-4-carboxaldehyde benzenesulfonate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.