missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methylpyrrole-d4 (ring-d4), CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | N-Methylpyrrole-d4 (ring-d4) |
| CAS | 190386-37-9 |
| Molecular Formula | C5 D4 H3 N |
| Quantity | 0.5 g |
| Synonym | 1 H-Pyrrole-2,3,4,5-d4, 1-methyl- (9 CI), 1-Methyl-1 H-pyrrole-2,3,4,5-d4, 1-Methylpyrrole-d4, N-Methylpyrrole-2,3,4,5-d4, N-Methylpyrrole-2,3,4,5-D4, N-Methylpyrrole-d4 (ring-d4) |
| InChI Formula | InChI=1 S/C5H7N/c1-6-4-2-3-5-6/h2-5 H,1H3/i2D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])n(C)c1[2 H] |
| IUPAC Name | 2,3,4,5-tetradeuterio-1-methylpyrrole |
| Molecular Weight (g/mol) | 85.14 |
| Formula Weight | 85.083 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction