missing translation for 'onlineSavingsMsg'
Learn More

N-Methylsarcosine, MedChemExpress

Product Code. 16492708
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
16492708 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 16492708

Brand: Medchem Express HYY0511

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress N-Methylsarcosine is an amino acid building block for protein, found in a small amount in the body.

N-Methylsarcosine is an amino acid building block for protein, found in a small amount in the body.

Specifications

Chemical Name or Material N-Methylsarcosine
CAS 1118-68-9
Color White
Purity Grade Notes Research
Molecular Formula C4H9NO2
Quantity 5 g
Synonym Dimethylglycine DMG N,N-Dimethylglycine
Solubility Information H2O : 100 mg/mL (969.74 mM; Need ultrasonic)
SMILES O=C(O)CN(C)C
Molecular Weight (g/mol) 103.12
Formula Weight 103.12
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.