missing translation for 'onlineSavingsMsg'
Learn More

N,N'-(1,2-Dihydroxyethylene)bisacrylamide, TRC

Product Code. 30387803
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387803 500 mg
30380903 100 mg
30380257 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387803 Supplier Toronto Research Chemicals Supplier No. TRCD493225500MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868-63-3
Molecular Formula C8H12N2O4
Molecular Weight (g/mol) 200.19
SMILES OC(NC(=O)C=C)C(O)NC(=O)C=C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H12N2O4/c1-3-5(11)9-7(13)8(14)10-6(12)4-2/h3-4,7-8,13-14H,1-2H2,(H,9,11)(H,10,12)
Formula Weight 200.0797
Chemical Name or Material N,N'-(1,2-Dihydroxyethylene)bisacrylamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.