missing translation for 'onlineSavingsMsg'
Learn More

N,N'-1,2-Ethanediylbis[N-(carboxymethyl)-glycine Tetrasodium Salt Hydrate, TRC

Product Code. 30381949
Change view
Click to view available options
Quantity:
1 g
50 g
Unit Size:
1g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30381949 1 g 1g
30370573 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381949 Supplier Toronto Research Chemicals Supplier No. TRCE9176601G
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N'-1,2-Ethanediylbis[N-(carboxymethyl)-glycine] Tetrasodium Salt Hydrate
CAS 194491-31-1
Molecular Formula C10H12N2Na4O8 • xH2O
Quantity 1 g
Synonym N,N’-1,2-Ethanediylbis[N-(Carboxymethyl)-Glycine Tetrasodium Salt Hydrate
InChI Formula InChI=1S/C10H16N2O8.4Na.H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;;;1H2/q;4*+1;/p-4
SMILES O=C(O[Na])CN(CC(O[Na])=O)CCN(CC(O[Na])=O)CC(O[Na])=O.[H2O]
IUPAC Name sodium 2,2',2'',2'''-(ethane-1,2-diylbis(azanetriyl))tetraacetate hydrate
Molecular Weight (g/mol) 398.19
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.