missing translation for 'onlineSavingsMsg'
Learn More

N,N,2,2-Tetramethyl-1,3-propanediamine, 97%, Thermo Scientific™

Product Code. 10863381 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10863381 100 g Glass bottle 100g
10351180 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10863381 Supplier Thermo Scientific Acros Supplier No. 210871000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Identificadores de productos químicos

CAS 53369-71-4
Molecular Formula C7H18N2
Molecular Weight (g/mol) 130.23
InChI Key ULDIVZQLPBUHAG-UHFFFAOYSA-N
Synonym n,n,2,2-tetramethyl-1,3-propanediamine, 1,3-propanediamine, n,n,2,2-tetramethyl, n,n,2,2-tetramethylpropane-1,3-diamine, n,n-dimethylneopentanediamine, 3-amino-2,2-dimethylpropyl dimethylamine, n1,n1,2,2-tetramethylpropane-1,3-diamine, 1,3-propanediamine, n1,n1,2,2-tetramethyl, acmc-209l6k, ksc597o9f, uldivzqlpbuhag-uhfffaoysa
PubChem CID 104477
IUPAC Name N',N',2,2-tetramethylpropane-1,3-diamine
SMILES CC(C)(CN)CN(C)C

Especificaciones

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8200g/mL
Boiling Point 155.0°C to 158.0°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2NCH2C(CH3)2CH2NH2
Refractive Index 1.4355
Quantity 100 g
Beilstein 04,III,596
Specific Gravity 0.82
Formula Weight 130.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N,N,2,2-Tetramethyl-1,3-propanediamine
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • N,N,2,2-Tetramethyl-1,3-propanediamine
Palabra de advertencia
  • Danger
Clasificación
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Indicaciones de peligro
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.