missing translation for 'onlineSavingsMsg'
Learn More

N,N-Bis(2-chloroethyl)-p-toluenesulfonamide, tech. 90%

Product Code. 11397375
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397375 5 g 5g
11307385 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397375 Supplier Thermo Scientific Alfa Aesar Supplier No. L05296.06
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 42137-88-2 | C11H15Cl2NO2S | 296.206 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42137-88-2
Molecular Formula C11H15Cl2NO2S
Molecular Weight (g/mol) 296.206
MDL Number MFCD00018944
InChI Key PTVBBIMKLOMGSY-UHFFFAOYSA-N
Synonym n,n-bis 2-chloroethyl-p-toluenesulfonamide, n,n-bis 2-chloroethyl-4-methylbenzenesulfonamide, n,n-bis 2-chloroethyl-p-toluenesulphonamide, n,n-bis .beta.-chloroethyl-p-toluenesulfonamide, n,n-bis 2-chloroethyl-4-toluenesulfonamide, n,n-bis 2'-chloroethyl-4-methylbenzenesulfonamide, n,n-bis 2-chloroethyl-4-methylbenzene sulfonamide, n,n-bis 2-chloroethyl-4-methylbenzene sulphonamide, benzenesulfonamide, n,n-bis 2-chloroethyl-4-methyl, n,n-bis-2-chloro-ethyl-4-methyl-benzenesulfonamide
PubChem CID 96039
IUPAC Name N,N-bis(2-chloroethyl)-4-methylbenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)N(CCCl)CCCl

Specifications

Melting Point 45°C to 47°C
Quantity 5 g
Beilstein 2134258
Formula Weight 296.22
Percent Purity 90%
Grade Technical
Chemical Name or Material N,N-Bis(2-chloroethyl)-p-toluenesulfonamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.