missing translation for 'onlineSavingsMsg'
Learn More

N.N'-Bis(2-hydroxyethyl)oxamide, 99%

Product Code. 11404437
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11404437 5 g 5g
11414437 25 g 25g
11424437 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404437 Supplier Thermo Scientific Alfa Aesar Supplier No. B20458.06
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1871-89-2 | C6H12N2O4 | 176.172 g/mol

N.N'-Bis(2-hydroxyethyl)oxamide is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N.N′-Bis(2-hydroxyethyl)oxamide is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1871-89-2
Molecular Formula C6H12N2O4
Molecular Weight (g/mol) 176.172
MDL Number MFCD00020559
InChI Key FPQJEXTVQZHURJ-UHFFFAOYSA-N
Synonym n,n'-bis 2-hydroxyethyl oxamide, n1,n2-bis 2-hydroxyethyl oxalamide, nn'-bis 2-hydroxyethyl oxamide, unii-t92n2yr78x, ethanediamide, n,n'-bis 2-hydroxyethyl, n,n-bis 2-hydroxyethyl oxamide, n,n'-bis-2-hydroxyethyl-oxamide, ethanediamide, n1,n2-bis 2-hydroxyethyl, n,n'-bis 2-hydroxyethyl ethanediamide, n-2-hydroxyethyl-n'-2-hydroxyethyl ethane-1,2-diamide
PubChem CID 74638
IUPAC Name N,N'-bis(2-hydroxyethyl)oxamide
SMILES C(CO)NC(=O)C(=O)NCCO

Specifications

Melting Point 167°C to 169°C (decomposition)
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 176.17
Percent Purity 99%
Chemical Name or Material N.N'-Bis(2-hydroxyethyl)oxamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.