missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N,N'-Bis(3-aminopropyl)-1,3-propanediamine, 97%

Product Code. 11345149
Click to view available options
Quantity:
5g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 11345149

Brand: Alfa Aesar™ A18133.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4605-14-5
Boiling Point 101°C to 103°C (1mmHg)
Flash Point >110°C (230°F)
Molecular Formula C9H24N4
MDL Number MFCD00008213
Quantity 5g
UN Number UN2735
Synonym norspermine, n,n'-bis 3-aminopropyl-1,3-propanediamine, 1,3-propanediamine, n,n'-bis 3-aminopropyl, n,n'-bis 3-aminopropyl propane-1,3-diamine, 1,5,9,13-tetraazatridecane, 3,3,3-tetramine, thermine, 1,3-propanediamine, n1,n3-bis 3-aminopropyl, n,n-bis 3-aminopropyl-1,3-propanediamine, n1,n1'-propane-1,3-diyl bis propane-1,3-diamine
InChI Key ZAXCZCOUDLENMH-UHFFFAOYSA-N
SMILES C(CN)CNCCCNCCCN
IUPAC Name N'-[3-(3-aminopropylamino)propyl]propane-1,3-diamine
Molecular Weight (g/mol) 188.319
PubChem CID 78350
ChEBI CHEBI:45718
Formula Weight 188.32
Percent Purity 97%
Chemical Name or Material N,N'-Bis(3-aminopropyl)-1,3-propanediamine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.