missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(3-aminopropyl)-1,3-propanediamine, 94%

Product Code. p-68193 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10332243

Brand: Thermo Scientific undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4605-14-5 | C9H24N4 | 188.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4605-14-5
Molecular Formula C9H24N4
Molecular Weight (g/mol) 188.32
MDL Number MFCD00008213
InChI Key ZAXCZCOUDLENMH-UHFFFAOYSA-N
Synonym norspermine, n,n'-bis 3-aminopropyl-1,3-propanediamine, 1,3-propanediamine, n,n'-bis 3-aminopropyl, n,n'-bis 3-aminopropyl propane-1,3-diamine, 1,5,9,13-tetraazatridecane, 3,3,3-tetramine, thermine, 1,3-propanediamine, n1,n3-bis 3-aminopropyl, n,n-bis 3-aminopropyl-1,3-propanediamine, n1,n1'-propane-1,3-diyl bis propane-1,3-diamine
PubChem CID 78350
ChEBI CHEBI:45718
IUPAC Name (3-aminopropyl)({3-[(3-aminopropyl)amino]propyl})amine
SMILES NCCCNCCCNCCCN

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 0.9200g/mL
Boiling Point 102°C (1.0 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula NH2(CH2)3NH(CH2)3NH(CH2)3NH2
Refractive Index 1.4900 to 1.492
Quantity 5 g
Specific Gravity 0.92
Formula Weight 188.32
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N, N'-Bis(3-aminopropyl)-1, 3-propanediamine, 97%
Show More Show Less
compliance-icons
Product Identifier
  • N,N'-Bis(3-aminopropyl)-1,3-propanediamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.