missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(trifluoroacetyl)-L-homocystine Dimethyl Ester, TRC

Product Code. 30335116
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335116 10 g 10g
30345235 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30335116 Supplier Toronto Research Chemicals Supplier No. TRCB58551010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N'-Bis(trifluoroacetyl)-L-homocystine Dimethyl Ester
CAS 84355-09-9
Molecular Formula C14H18F6N2O6S2
Quantity 10 g
Synonym 4,4'-Dithiobis[2-[(trifluoroacetyl)amino]-butanoic Acid Dimethyl Ester, (2S,2'S)-4,4'-Dithiobis[2-[(2,2,2-trifluoroacetyl)amino]-butanoic Acid 1,1'-Dimethyl Ester
InChI Formula InChI=1S/C14H18F6N2O6S2/c1-27-9(23)7(21-11(25)13(15,16)17)3-5-29-30-6-4-8(10(24)28-2)22-12(26)14(18,19)20/h7-8H,3-6H2,1-2H3,(H,21,25)(H,22,26)/t7-,8-/m0/s1
SMILES COC(=O)[C@H](CCSSCC[C@H](NC(=O)C(F)(F)F)C(=O)OC)NC(=O)C(F)(F)F
IUPAC Name methyl (2S)-4-[[(3S)-4-methoxy-4-oxo-3-[(2,2,2-trifluoroacetyl)amino]butyl]disulfanyl]-2-[(2,2,2-trifluoroacetyl)amino]butanoate
Molecular Weight (g/mol) 488.42
Formula Weight 488.051
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.