missing translation for 'onlineSavingsMsg'
Learn More

N-(n-Butyl)aniline, 99%

Product Code. 11421217
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11421217 100 g 100g
11411217 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11421217 Supplier Thermo Scientific Alfa Aesar Supplier No. B24180.22
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1126-78-9 | C10H15N | 149.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1126-78-9
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00035798
InChI Key VSHTWPWTCXQLQN-UHFFFAOYSA-N
Synonym benzenamine, n-butyl, aniline, n-butyl, 4-phenylamino butane, n-butylbenzenamine, n-n-butyl aniline, butylaniline, n-phenyl-n-butylamine, butylphenylamine, unii-r2znj7l2um, benzamine, n-butyl
PubChem CID 14310
IUPAC Name N-butylaniline
SMILES CCCCNC1=CC=CC=C1

Specifications

Density 0.932
Boiling Point 240°C to 242°C
Flash Point 105°C (222°F)
Refractive Index 1.532
Quantity 100 g
UN Number UN2738
Formula Weight 149.23
Percent Purity 99%
Chemical Name or Material N-(n-Butyl)aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.