missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di(1-naphthyl)-N,N'-diphenylbenzidine, 98%, Thermo Scientific Chemicals

Product Code. 15478956
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15478956 250 mg 250mg
15411749 1 g 1g
15421749 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15478956 Supplier Thermo Scientific Alfa Aesar Supplier No. H64502.MD
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123847-85-8
Molecular Formula C44H32N2
Molecular Weight (g/mol) 588.754
MDL Number MFCD03093246
InChI Key IBHBKWKFFTZAHE-UHFFFAOYSA-N
Synonym n4,n4'-di naphthalen-1-yl-n4,n4'-diphenyl-1,1'-biphenyl-4,4'-diamine, alpha-npb, n,n'-bis naphthalen-1-yl-n,n'-bis phenyl benzidine, 4,4'-bis n-1-naphthyl-n-phenylamino biphenyl, n,n'-di 1-naphthyl-n,n'-diphenylbenzidine, n,n'-bis-1-naphthalenyl-n,n'-bis-phenyl-1,1'-biphenyl-4,4'-diamine, 1,1'-biphenyl-4,4'-diamine, n,n'-di-1-naphthalenyl-n,n'-diphenyl, n,n'-di-1-naphthyl-n,n'-diphenylbenzidine
PubChem CID 5069127
IUPAC Name N-[4-[4-(N-naphthalen-1-ylanilino)phenyl]phenyl]-N-phenylnaphthalen-1-amine
SMILES C1=CC=C(C=C1)N(C2=CC=C(C=C2)C3=CC=C(C=C3)N(C4=CC=CC=C4)C5=CC=CC6=CC=CC=C65)C7=CC=CC8=CC=CC=C87

Specifications

Melting Point 279°C to 283°C
Color Yellow
Quantity 250 mg
Beilstein 8022378
Formula Weight 588.75
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N,N'-Di(1-naphthyl)-N,N'-diphenylbenzidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.