missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di-Boc-1H-pyrazole-1-carboxamidine, 98+%

Product Code. 15480828
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15480828 5 g 5g
15470828 1 g 1g
15490828 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15480828 Supplier Thermo Scientific Alfa Aesar Supplier No. H54565.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 152120-54-2 | C14H22N4O4 | 310.354 g/mol

N,N'-Di-Boc-1H-pyrazole-1-carboxamidine is used in the stereoselective synthesis of the bicyclic guanidine alkaloid (+)-monanchorin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Di-Boc-1H-pyrazole-1-carboxamidine is used in the stereoselective synthesis of the bicyclic guanidine alkaloid (+)-monanchorin.

Solubility
Slightly soluble in water. Soluble in methanol, and chloroform.

Notes
Store at 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152120-54-2
Molecular Formula C14H22N4O4
Molecular Weight (g/mol) 310.354
MDL Number MFCD01075122
InChI Key QFNFDHNZVTWZED-UHFFFAOYSA-N
Synonym n,n'-di-boc-1h-pyrazole-1-carboxamidine, bis-boc-pyrazolocarboxamidine, pyrazol boc 2, n,n-bis-boc-1-guanylpyrazole, n,n'-bis-boc-1-guanylpyrazol, n,n'-bis-boc-1-guanylpyrazole, n,n'-bisboc-pyrazole-1-carboximidamide, tert-butyl tert-butoxycarbonyl amino 1h-pyrazol-1-yl methylene carbamate, tert-butyl boc-amino 1h-pyrazol-1-yl methylene carbamate, 1-n,n'-di-boc amidino pyrazole
PubChem CID 6383521
IUPAC Name tert-butyl (NZ)-N-[[(2-methylpropan-2-yl)oxycarbonylamino]-pyrazol-1-ylmethylidene]carbamate
SMILES CC(C)(C)OC(=O)NC(=NC(=O)OC(C)(C)C)N1C=CC=N1

Specifications

Melting Point 86°C to 90°C
Quantity 5 g
Solubility Information Slightly soluble in water. Soluble in methanol,and chloroform.
Formula Weight 310.35
Percent Purity ≥98%
Chemical Name or Material N,N'-Di-Boc-1H-pyrazole-1-carboxamidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.