missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di-BOC-1H-pyrazole-1-carboxamidine, 98%

Product Code. 10316744 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10316744 1 g 1g
10744385 5 g 5g
10604185 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10316744 Supplier Thermo Scientific Acros Supplier No. 427970010
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 152120-54-2 | C14H22N4O4 | 310.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152120-54-2
Molecular Formula C14H22N4O4
Molecular Weight (g/mol) 310.35
InChI Key QFNFDHNZVTWZED-UHFFFAOYSA-N
Synonym n,n'-di-boc-1h-pyrazole-1-carboxamidine, bis-boc-pyrazolocarboxamidine, pyrazol boc 2, n,n-bis-boc-1-guanylpyrazole, n,n'-bis-boc-1-guanylpyrazol, n,n'-bis-boc-1-guanylpyrazole, n,n'-bisboc-pyrazole-1-carboximidamide, tert-butyl tert-butoxycarbonyl amino 1h-pyrazol-1-yl methylene carbamate, tert-butyl boc-amino 1h-pyrazol-1-yl methylene carbamate, 1-n,n'-di-boc amidino pyrazole
PubChem CID 6383521
IUPAC Name tert-butyl (NZ)-N-[[(2-methylpropan-2-yl)oxycarbonylamino]-pyrazol-1-ylmethylidene]carbamate
SMILES CC(C)(C)OC(=O)NC(=NC(=O)OC(C)(C)C)N1C=CC=N1

Specifications

Melting Point 86.0°C to 90.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 310.35
Percent Purity 98%
Chemical Name or Material N, N'-Di-BOC-1H-pyrazole-1-carboxamidine
compliance-icons
Product Identifier
  • N,N'-Di-BOC-1H-pyrazole-1-carboxamidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.