missing translation for 'onlineSavingsMsg'
Learn More

N,N-Di-n-butylformamide, 99%

Product Code. 11343356
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343356 25 g 25g
11353356 100 g 100g
11363356 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11343356 Supplier Thermo Scientific Alfa Aesar Supplier No. L12755.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 761-65-9 | C9H19NO | 157.26 g/mol

N,N-Di-n-butylformamide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Di-n-butylformamide is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 761-65-9
Molecular Formula C9H19NO
Molecular Weight (g/mol) 157.26
MDL Number MFCD00003289
InChI Key NZMAJUHVSZBJHL-UHFFFAOYSA-N
Synonym dibutylformamide, formamide, n,n-dibutyl, n,n-di-n-butylformamide, formamide, n,n-di-n-butyl, dibutylamid kyseliny mravenci, dibutylamid kyseliny mravenci czech, formamide, dibutyl, n-formyldibutylamine, formamide,n-dibutyl, n,n-di-butylformamide
PubChem CID 12975
IUPAC Name N,N-dibutylformamide
SMILES CCCCN(CCCC)C=O

Specifications

Melting Point <-30°C
Density 0.864
Boiling Point 119°C to 120°C (15 mmHg)
Flash Point 115°C (239°F)
Refractive Index 1.4445
Quantity 25 g
Beilstein 636100
Solubility Information Insoluble in water.
Formula Weight 157.26
Percent Purity 99%
Chemical Name or Material N,N-Di-n-butylformamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.