missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diacetylethylenediamine, 98%, Thermo Scientific™

Product Code. 11463834
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463834 50 g 50g
11453834 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11463834 Supplier Thermo Scientific Alfa Aesar Supplier No. A11866.18
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871-78-3
Molecular Formula C6H12N2O2
Molecular Weight (g/mol) 144.174
MDL Number MFCD00014966
InChI Key WNYIBZHOMJZDKN-UHFFFAOYSA-N
Synonym n,n'-diacetylethylenediamine, n,n'-ethylenebisacetamide, n-2-acetamidoethyl acetamide, unii-sxn3k5812f, acetamide, n,n'-1,2-ethanediylbis, nn'-diacetylethylenediamine, n-2-acetylamino ethyl acetamide, n,n'-ethylenedi diacetamide, diacetylethylenediamine, acetamide,n'-ethylenebis
PubChem CID 70097
IUPAC Name N-(2-acetamidoethyl)acetamide
SMILES CC(=O)NCCNC(=O)C

Specifications

Melting Point 172°C to 176°C
Quantity 50 g
Beilstein 1762229
Formula Weight 144.17
Percent Purity 98%
Chemical Name or Material N,N'-Diacetylethylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.