missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diacetylhydrazine, 98%

Product Code. 11495393
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495393 250 g 250g
11475393 10 g 10g
11485393 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495393 Supplier Thermo Scientific Alfa Aesar Supplier No. A13393.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3148-73-0 | C4H8N2O2 | 116.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3148-73-0
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00008673
InChI Key ZLHNYIHIHQEHJQ-UHFFFAOYSA-N
Synonym n,n'-diacetylhydrazine, 1,2-diacetylhydrazine, acetic acid, 2-acetylhydrazide, diacetylahydrazine, diacetylhydrazine, sym-diacetylhydrazine, unii-ymr3zf0q4m, hydrazine, 1,2-diacetyl, 1.2-diacetylhydrazine, ymr3zf0q4m
PubChem CID 72884
ChEBI CHEBI:80616
IUPAC Name N'-acetylacetohydrazide
SMILES CC(=O)NNC(=O)C

Specifications

Melting Point 138°C to 142°C
Boiling Point 209°C (15 mmHg)
Quantity 250 g
Beilstein 1754018
Formula Weight 116.12
Percent Purity 98%
Chemical Name or Material N,N'-Diacetylhydrazine
compliance-icons
Product Identifier
  • N,N'-Diacetylhydrazine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Acetic acid, 2-acetylhydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.