missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diallyl-L-tartardiamide, 99%

Product Code. 11444853
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444853 250 g 250g
11434853 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444853 Supplier Thermo Scientific Alfa Aesar Supplier No. A12195.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 58477-85-3 | C10H16N2O4 | 228.25 g/mol

Cross-linking co-monomer for the polymerization of soluble polyacrylamide gels

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58477-85-3
Molecular Formula C10H16N2O4
Molecular Weight (g/mol) 228.25
MDL Number MFCD00008640
InChI Key ZRKLEAHGBNDKHM-OCAPTIKFSA-N
Synonym n,n'-diallyltartardiamide, 2r,3s-2,3-dihydroxy-n,n'-bis prop-2-enyl butanediamide, 2r,3s-2,3-dihydroxy-n,n'-bis prop-2-en-1-yl succinamide
PubChem CID 6994946
IUPAC Name (2S,3S)-2,3-dihydroxy-N,N'-bis(prop-2-enyl)butanediamide
SMILES O[C@@H]([C@@H](O)C(=O)NCC=C)C(=O)NCC=C

Specifications

Melting Point 184°C to 186°C
Quantity 250 g
Beilstein 1712934
Optical Rotation +118° (c=3 in methanol)
Formula Weight 228.25
Percent Purity 99%
Chemical Name or Material N,N'-Diallyl-L-tartardiamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.