missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diallyl-L-tartardiamide, Electrophoresis Reagent, 99%, Thermo Scientific™

Product Code. 15410617
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15410617 250 g 250g
15400617 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15410617 Supplier Thermo Scientific Alfa Aesar Supplier No. J66587.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58477-85-3
Molecular Formula C10H16N2O4
Molecular Weight (g/mol) 228.25
MDL Number MFCD00008640
InChI Key ZRKLEAHGBNDKHM-OCAPTIKFSA-N
Synonym n,n'-diallyltartardiamide, 2r,3s-2,3-dihydroxy-n,n'-bis prop-2-enyl butanediamide, 2r,3s-2,3-dihydroxy-n,n'-bis prop-2-en-1-yl succinamide
PubChem CID 6994946
SMILES O[C@@H]([C@@H](O)C(=O)NCC=C)C(=O)NCC=C

Specifications

Melting Point 184°C to 186°C
Color White
Quantity 250 g
Beilstein 1712934
IUPAC Name (2R,3S)-2,3-dihydroxy-N,N'-bis(prop-2-en-1-yl)butanediamide
Formula Weight 228.25
Percent Purity 99%
Physical Form Flakes
Chemical Name or Material N,N'-Diallyl-L-tartardiamide, Electrophoresis Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.