missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diaminoguanidine monohydrochloride, 98%

Product Code. 11415217
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11415217 100 g 100g
11495207 5 g 5g
11405217 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11415217 Supplier Thermo Scientific Alfa Aesar Supplier No. B22615.22
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 36062-19-8 | CH8ClN5 | 125.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36062-19-8
Molecular Formula CH8ClN5
Molecular Weight (g/mol) 125.56
MDL Number MFCD00012948
InChI Key HAZRIBSLCUYMQP-UHFFFAOYSA-N
Synonym 1,3-diaminoguanidine hydrochloride, unii-014y506oeg, 1,3-diaminoguanidine monohydrochloride, carbonimidic dihydrazide, hydrochloride, n,n'-diaminoguanidine monohydrochloride, n,n'-diaminoguanidine, carbonimidic dihydrazide, hydrochloride 1:?, 1,2-diaminoguanidine hydrochloride, n,n'-diaminoguandine, hydrochloride, pubchem20888
PubChem CID 9566041
IUPAC Name 1,2-diaminoguanidine;hydrochloride
SMILES C(=NN)(N)NN.Cl

Specifications

Melting Point ∼181°C (decomposition)
Quantity 100 g
Beilstein 3556702
Sensitivity Hygroscopic
Formula Weight 125.56
Percent Purity 98%
Chemical Name or Material N,N'-Diaminoguanidine monohydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.